CAS RN 34598-33-9
IUPAC Navn:
4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heptadecafluoroundecanoic acid
Synonymer:
4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heptadecafluoroundecanoic acid
Molekylvægt (g/mol):
492.13
Molekylformel:
C11H5F17O2
InChi Key:
JZRCRCFPVAXHHQ-UHFFFAOYSA-N
SMILES:
OC(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
1
–
2
af
2
Resultater
1
Kampagner er tilgængelige
2H,2H,3H,3H-Perfluoroundecanoic acid 50 μg/mL in Methanol, Dr. Ehrenstorfer
2H,2H,3H,3H-Perfluoroundecanoic acid 50 μg/mL in Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecanoic Acid, TRC
CAS: 34598-33-9 Molekylær formel: C11 H5 F17 O2 Molekylvægt (g/mol): 492.13 Synonym: 4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecanoic acid,2H,2H,3H,3H-Perfluoroundecanoic acid,3-Perfluorooctylpropionic acid,Heptadecafluoroundecanoic acid IUPAC navn: 4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heptadecafluoroundecanoic acid SMIL: OC(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F