CAS RN 33838-52-7
IUPAC Navn:
heptane
Synonymer:
heptane N-heptane eptani skellysolve C dipropyl methane gettysolve-C hsdb 90
Molekylvægt (g/mol):
100.21
Molekylformel:
C7H16
InChi Key:
IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES:
CCCCCCC
1
–
2
af
2
Resultater
2
n-Heptane-d16 (Reagent grade), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.