CAS RN 33311-76-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
2-(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)-N-[4-nitro-2-(phenylcarbonyl)-phenyl]acetamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.