CAS RN 33297-48-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
N-Acetyl-S-(2-ethoxycarbonylethyl-1-methyl)-L-cysteine, Dicyclohexylammonium Salt(Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-Acetyl-S-(2-ethoxycarbonylethyl-1-methyl)-L-cysteine, Dicyclohexylammonium Salt(Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-Acetyl-S-(2-ethoxycarbonylethyl-1-methyl)-L-cysteine, Dicyclohexylammonium Salt(Mixture of Diastereomers), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.