CAS RN 30363-03-2
IUPAC Navn:
tris(4-bromophenyl)-1,3,5-triazine
Synonymer:
tris(4-bromophenyl)-1,3,5-triazine
Molekylvægt (g/mol):
546.06
Molekylformel:
C21H12Br3N3
InChi Key:
WZYVDGDZBNQVCF-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1
1
–
3
af
3
Resultater
1
2,4,6-Tris-(4-bromophenyl)[1,3,5]triazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,4,6-Tris-(4-bromophenyl)[1,3,5]triazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,4,6-Tris-(4-bromophenyl)[1,3,5]triazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.