CAS RN 30189-85-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
PPA-904, MedChemExpress
MedChemExpress PPA-904 is a specific phenothiazine photosensitizer in photodynamic therapy (PDT) research, especially topical application for cutaneous leishmaniasis in vivo.