Få mere at vide
PPA-904, MedChemExpress
Artikelnummer. 16432388
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
10 mg
20 mg
Pakningsstørrelse:
1mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16432388 5 mg -
16473169 1 mg 1mg
16430189 10 mg -
16420706 20 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16432388 Leverandør Medchem Express Leverandørnr. HYU00128
MedChemExpress PPA-904 is a specific phenothiazine photosensitizer in photodynamic therapy (PDT) research, especially topical application for cutaneous leishmaniasis in vivo.
PPA-904 is a specific phenothiazine photosensitizer in photodynamic therapy (PDT) research, especially topical application for cutaneous leishmaniasis in vivo.
Tekniske data
Kemisk navn eller materiale PPA-904
CAS 30189-85-6
Farve Coffee
Renhedsgrad noter Research
Molekylær formel C28H42BrN3S
Mængde 5 mg
SMIL CCCCN(C1=CC2=[S+]C3=C(C=CC(N(CCCC)CCCC)=C3)N=C2C=C1)CCCC.[Br-]
Molekylvægt (g/mol) 532.62
Formel vægt 532.62
Procent renhed 98.12%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation