CAS RN 301353-96-8
IUPAC Navn:
1-(3,6-dibromo-9H-carbazol-9-yl)-3-(phenylamino)propan-2-ol
Synonymer:
1-(3,6-dibromocarbazol-9-yl)-3-(phenylamino)propan-2-ol
Molekylvægt (g/mol):
474.2
Molekylformel:
C21H18Br2N2O
InChi Key:
FZHHRERIIVOATI-UHFFFAOYNA-N
SMILES:
OC(CNC1=CC=CC=C1)CN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2
1
–
2
af
2
Resultater
1
Tocris Bioscience™ P7C3
NAMPT Activators
2
P7C3, MedChemExpress
MedChemExpress P7C3 is an orally bioavailable and blood-brain barrier penetrant aminopropyl carbazole, with neuroprotective effects. P7C3 can be used for the research of neurodegenerative diseases, including Parkinson's disease.