CAS RN 3007-75-8
IUPAC Navn:
N,N-dioctylaniline
Synonymer:
N,N-dioctylaniline
Molekylvægt (g/mol):
317.56
Molekylformel:
C22H39N
InChi Key:
PTRSTXBRQVXIEW-UHFFFAOYSA-N
SMILES:
CCCCCCCCN(CCCCCCCC)C1=CC=CC=C1
1
–
1
af
1
Resultater
1
N,N-Di-n-octylanilin, 98%, Thermo Scientific Chemicals
CAS: 3007-75-8 Molekylær formel: C22H39N Molekylvægt (g/mol): 317.56 MDL nummer: MFCD00155305 InChI nøgle: PTRSTXBRQVXIEW-UHFFFAOYSA-N Synonym: n,n-di-n-octylaniline,benzenamine, n,n-dioctyl,dioctylphenylamine,benzenamine,n,n-dioctyl PubChem CID: 137791 IUPAC navn: N,N-dioktylanilin SMIL: CCCCCCCCN(CCCCCCCC)C1=CC=CC=C1