CAS RN 298679-54-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
7-[[4-[Bis(3-amino-3-oxopropyl)amino]-6-chloro-1,3,5-triazin-2-yl]amino]-4-hydroxy-3-[2-(4-methoxyphenyl)diazenyl]-2-naphthalenesulfonic Acid 1-Methylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
7-[[4-[Bis(3-amino-3-oxopropyl)amino]-6-chloro-1,3,5-triazin-2-yl]amino]-4-hydroxy-3-[2-(4-methoxyphenyl)diazenyl]-2-naphthalenesulfonic Acid 1-Methylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
7-[[4-[Bis(3-amino-3-oxopropyl)amino]-6-chloro-1,3,5-triazin-2-yl]amino]-4-hydroxy-3-[2-(4-methoxyphenyl)diazenyl]-2-naphthalenesulfonic Acid 1-Methylethyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.