CAS RN 290304-24-4
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Daun02, MedChemExpress
MedChemExpress Daun02 is a prodrug of the topoisomerase inhibitor Daunorubicin.