CAS RN 27867-02-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
trans-1,2,3,4,4a,10,11,11a-Octahydro-N,N-dimethyl-5H-Dibenzo[a,d]cycloheptene-Delta5,Gamma-propylamine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
trans-1,2,3,4,4a,10,11,11a-Octahydro-N,N-dimethyl-5H-Dibenzo[a,d]cycloheptene-Delta5,Gamma-propylami, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.