CAS RN 2755075-91-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ JNJ 1013NC
Negative control for the IRAK1 degrader JNJ 1013 (Cat. No. 8029)