CAS RN 2724726-76-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
3-Methyl 5-[Methyl(phenylmethyl)amino]ethyl 2,6-Dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate Dihydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.