CAS RN 2724511-24-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
4-(Ethylmethylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate Hydrochloride (Methylethyl Analogue of Oxybutynin Hydrochloride), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.