Få mere at vide
4-(Ethylmethylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate Hydrochloride (Methylethyl Analogue of Oxybutynin Hydrochloride), Mikromol™
Beskrivelse
4-(Ethylmethylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate Hydrochloride (Methylethyl Analogue of Oxybutynin Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-(Ethylmethylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate Hydrochloride |
| CAS | 2724511-24-2 |
| Analyt- eller komponentnavne | 4-(Ethylmethylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H29 N O3 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 4-(Ethylmethylamino)but-2-ynyl (RS)-2-Cyclohexyl-2-hydroxy-2-phenylacetate Hydrochloride, Oxybutynin Hydrochloride Imp. C (EP) as Hydrochloride, Methylethyl Analogue of Oxybutynin Hydrochloride, Oxybutynin Related Compound C as Hydrochloride, Oxybutynin Hydrochloride Impurity C as Hydrochloride, Oxybutynin Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C21H29NO3.ClH/c1-3-22(2)16-10-11-17-25-20(23)21(24,18-12-6-4-7-13-18)19-14-8-5-9-15-19;/h4,6-7,12-13,19,24H,3,5,8-9,14-17H2,1-2H3;1H |
| SMIL | Cl.CCN(C)CC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2 |
| IUPAC navn | 4-[ethyl(methyl)amino]but-2-ynyl 2-cyclohexyl-2-hydroxy-2-phenylacetate;hydrochloride |
| Vis mere |