CAS RN 2717331-30-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
4-Chloro Mepivacaine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(RS)-N-(4-Chloro-2,6-dimethylphenyl)-1-methylpiperidine-2-carboxamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
4-Chloro Mepivacaine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.