CAS RN 2442597-56-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ VU 6015929
Potent and selective discoidin domain receptor 1/2 inhibitor