CAS RN 244-76-8
IUPAC Navn:
9H-pyrido[2,3-b]indole
Synonymer:
9H-pyrido[2,3-b]indole
Molekylvægt (g/mol):
168.2
Molekylformel:
C11H8N2
InChi Key:
BPMFPOGUJAAYHL-UHFFFAOYSA-N
SMILES:
N1C2=C(C=CC=C2)C2=C1N=CC=C2
1
–
1
af
1
Resultater
1
alpha-Carboline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.