CAS RN 24332-20-5
IUPAC Navn:
1,1,2-trimethoxyethane
Synonymer:
bis-methoxyethoxy methane
Molekylvægt (g/mol):
120.15
Molekylformel:
C5H12O3
InChi Key:
DYOZNCVZPFIXLU-UHFFFAOYSA-N
SMILES:
COCC(OC)OC
1
–
3
af
3
Resultater
1
Methoxyacetaldehyde Dimethyl Acetal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Methoxyacetaldehyde Dimethyl Acetal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Methoxyacetaldehyde Dimethyl Acetal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.