CAS RN 24307-26-4
IUPAC Navn:
1,1-dimethylpiperidin-1-ium chloride
Synonymer:
mepiquat chloride
Molekylvægt (g/mol):
149.66
Molekylformel:
C7H16ClN
InChi Key:
VHOVSQVSAAQANU-UHFFFAOYSA-M
SMILES:
[Cl-].C[N+]1(C)CCCCC1
1
–
3
af
3
Resultater
1
Mepiquat Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Mepiquat Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Mepiquat Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.