missing translation for 'onlineSavingsMsg'
Learn More

Mepiquat Chloride, TRC

Product Code. 30464581
Change view
Click to view available options
Mængde:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30464581 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464581 Supplier Toronto Research Chemicals Supplier No. TRCM2250905G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Mepiquat chloride
CAS 24307-26-4
Molekylær formel C7 H16 N . Cl
Mængde 5 g
Synonym 1,1-Dimethylpiperidinium chloride (7CI), Piperidinium, 1,1-dimethyl-, chloride (8CI,9CI), BAS 083, BAS 08301W, BAS 85559X, CDP, Dimethylpiperidinium chloride, Megahix, Mepex, Mepichlor, Mepiquat chloride, N,N-Dimethylpiperidinium chloride, Pix, Pix Ultra
InChI formel InChI=1S/C7H16N.ClH/c1-8(2)6-4-3-5-7-8;/h3-7H2,1-2H3;1H/q+1;/p-1
SMIL [Cl-].C[N+]1(C)CCCCC1
IUPAC navn 1,1-dimethylpiperidin-1-ium;chloride
Molekylvægt (g/mol) 149.66
Formel vægt 149.0971
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.