CAS RN 2429-73-4
IUPAC Navn:
trisodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-7-amino-1-oxo-3-sulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
Synonymer:
trisodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-7-amino-1-oxo-3-sulfonatonaphthalen-2-ylidene]hydrazin-1-yl}-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxonaphthalene-2,7-disulfonate
Molekylvægt (g/mol):
830.7
Molekylformel:
C32H21N6Na3O11S3
InChi Key:
ZAOMBAUENFURGO-OWOLPYOXSA-K
SMILES:
[Na+].[Na+].[Na+].NC1=CC=C2C=C(\C(=N/NC3=CC=C(C=C3)C3=CC=C(N\N=C4\C(=O)C5=C(N)C=C(C=C5C=C4S([O-])(=O)=O)S([O-])(=O)=O)C=C3)C(=O)C2=C1)S([O-])(=O)=O
1
–
3
af
3
Resultater
1
Direct Blue 2 (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Direct Blue 2 (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Direct Blue 2 (Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.