missing translation for 'onlineSavingsMsg'
Få mere at vide

Direct Blue 2 (Technical Grade), TRC

Artikelnummer. 30374009
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30374009 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30374009 Leverandør Toronto Research Chemicals Leverandørnr. TRCD494985250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Direct Blue 2 (Technical Grade)
CAS 2429-73-4
Molekylær formel C32H21N6Na3O11S3
Mængde 250 mg
Synonym 2,7-Naphthalenedisulfonic acid, 5-amino-3-[2-[4'-[2-(7-amino-1-hydroxy-3-sulfo-2-naphthalenyl)diazenyl][1,1'-biphenyl]-4-yl]diazenyl]-4-hydroxy-, sodium salt (1:3), 2,7-Naphthalenedisulfonic acid, 5-amino-3-[[4'-[(7-amino-1-hydroxy-3-sulfo-2-naphthalenyl)azo][1,1'-biphenyl]-4-yl]azo]-4-hydroxy-, trisodium salt (9CI), C.I. Direct Blue 2 (7CI), C.I. Direct Blue 2, trisodium salt (8CI), Direct Diazo Black S (6CI), Airedale Black BHD, Aizen Direct Black BH, Altazine Black BH, Amanil Developed Black BHSW, Atul Developed Black BT, Azine Diazo Black BHK, Azocard Blue BH, Azomine Black BH, Belamine Diazo Black BH, Bencidal Navy Blue BH, Benzanil Black BH, Benzo Black Blue BH, Benzo Black Blue FBH, Blue BH, Brasilazol Black BH, C.I. 22590, Calcoloid Diazo Black BHL, Calcomine Diazo Black BHD, Calcomine Diazo Black BTCW, Chloramine Black BH, Chlorazol Black BH, Chlorazol Leather Black BH, Chrome Leather Black BH, Chrome Leather Black CR, Chrome Leather Black DS, Chrome Leather Dark Blue BHM, Cutamin Dark Blue CB, Diacotton Black BH, Diamine Black BH, Diamine Black BHM, Diaminogene Velour Black B, Dianil Dark Blue H, Diaphtamine Black BH, Diazine Black BHC, Diazine Black H, Diazine Black HDW, Diazine Black HNJ, Diazo Black BH, Diazo Black BHN-CF, Diazo Black BHSW, Diazo Black BHSWK, Diazo Black CR, Diazo Direct Black N, Diazo Fast Black BH, Diazo Fast Black MBH, Diazo Navy Blue BH, Diazol Black BH, Diazophenyl Black BH, Diphenyl Blue Black GHS, Diphenyl Blue Black MBH, Direct Black BH, Direct Blue Black BH, Direct Dark Blue BH, Direct Diazo Black, Direct Diazo Black C, Direct Diazo Black N, Direct Navy Blue BH, Eniazol Blue Black BHN, Fenamin Navy Blue H, Fixanol Blue BH, Indoxine KL, Japanol Black BHK, Kayaku Direct Black BH, Melantherine BH, Melantherine BHX, Mitsui Direct Black BH, Navy Blue EMBL, Neklamin Black BH, Orbamin Black BH, Paramine Black BH, Phenazo Black BH, Pheno Navy Blue, Pontamine Deep Blue BH, Pontamine Diazo Black BHSW, Symulon Direct Black BH, Tertrodirect Black BH, Tertrodirect Black BHS, Union Fast Navy Blue DS, Vondacel Dark Blue BH, Zambesi Dark Blue BH
InChI formel InChI=1S/C32H24N6O11S3.3Na/c33-20-6-1-18-12-26(51(44,45)46)29(31(39)24(18)14-20)37-35-21-7-2-16(3-8-21)17-4-9-22(10-5-17)36-38-30-27(52(47,48)49)13-19-11-23(50(41,42)43)15-25(34)28(19)32(30)40;;;/h1-15,39-40H,33-34H2,(H,41,42,43)(H,44,45,46)(H,47,48,49);;;/q;3*+1/p-3
SMIL [Na+].[Na+].[Na+].Nc1ccc2cc(c(N=Nc3ccc(cc3)c4ccc(cc4)N=Nc5c(O)c6c(N)cc(cc6cc5S(=O)(=O)[O-])S(=O)(=O)[O-])c(O)c2c1)S(=O)(=O)[O-]
IUPAC navn trisodium;5-amino-3-[(E)-[4-[4-[(E)-(7-amino-1-hydroxy-3-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]-4-hydroxynaphthalene-2,7-disulfonate
Molekylvægt (g/mol) 830.71
Formel vægt 830.0124
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.