CAS RN 2411098-96-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ Thalidomide 4'-ether-alkylC5-azide
Cereblon ligand with alkyl linker and terminal azide for onward chemistry