missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Thalidomide 4'-ether-alkylC5-azide

Description
Thalidomide 4'-ether-alkylC5-azide is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus an alkylC5 linker with a terminal azide ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Kemisk navn eller materiale | 4-(5-Azidopentoxy)-2-(2,6-dioxopiperidin-3 yl)-1 H-isoindole-1,3(2 H)-dione |
| CAS | 2411098-96-7 |
| Mængde | 25 mg |
| Mål | Degrader Building Blocks |
| Molekylær formel | C18H19N5O5 |
| Renhed | 0.95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?