CAS RN 2408341-97-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ VH 101, acid
VHL ligand, carboxylic acid functionalized at the benzylic position