CAS RN 24047-72-1
IUPAC Navn:
2-hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one
Synonymer:
oxypinocamphone
Molekylvægt (g/mol):
168.24
Molekylformel:
C10H16O2
InChi Key:
VZRRCQOUNSHSGB-UHFFFAOYNA-N
SMILES:
CC1(C)C2CC1C(C)(O)C(=O)C2
1
–
1
af
1
Resultater
1
(+)-(1R,2R,5R)-2-Hydroxy-3-pinanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.