CAS RN 23491-54-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
MedChemExpress Hoechst 33258 analog 2
MedChemExpress Hoechst 33258 analog 2 are part of a family of blue fluorescent dyes used to stain DNA.