CAS RN 23454-33-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Trenbolone cyclohexylmethylcarbonate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Trenbolone Cyclohexylmethylcarbonate (Trenbolone Hexahydrobenzylcarbonate), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Trenbolone Cyclohexylmethylcarbonate (Trenbolone Hexahydrobenzylcarbonate), LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.