CAS RN 23422-53-9
IUPAC Navn:
hydrogen 3-{[(dimethylamino)methylidene]amino}phenyl N-methylcarbamate chloride
Synonymer:
formetanate
Molekylvægt (g/mol):
257.72
Molekylformel:
C11H16ClN3O2
InChi Key:
MYPKGPZHHQEODQ-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CNC(=O)OC1=CC=CC(=C1)N=CN(C)C
1
–
3
af
3
Resultater
1
Formetanate Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Formetanate Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Formetanate Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.