CAS RN 2341841-08-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Tocris Bioscience™ uPSEM 792 hydrochloride
Highly potent PSAM4-GlyR and PSAM4-5-HT3 chimeric ion channel agonist; brain-penetrant