CAS RN 2341-22-2
IUPAC Navn:
4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-fluoro-1,2-dihydropyrimidin-2-one
Synonymer:
5-fluorocytidine
Molekylvægt (g/mol):
261.21
Molekylformel:
C9H12FN3O5
InChi Key:
STRZQWQNZQMHQR-UAKXSSHOSA-N
SMILES:
NC1=NC(=O)N(C=C1F)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
1
–
2
af
2
Resultater
1
5-Fluoro Cytidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Fluoro Cytidine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.