CAS RN 23202-66-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
3
af
3
Resultater
1
Bromo 2,4,6-Tri-O-acetyl-3-O-(2,3,4,6-tetra-O-acetyl -b-D-glucopyranosyl)-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Bromo 2,4,6-Tri-O-acetyl-3-O-(2,3,4,6-tetra-O-acetyl -b-D-glucopyranosyl)-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Bromo 2,4,6-Tri-O-acetyl-3-O-(2,3,4,6-tetra-O-acetyl -b-D-glucopyranosyl)-Alpha-D-glucopyranoside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.