CAS RN 229625-50-7
IUPAC Navn:
di-tert-butyl chloromethyl phosphate
Synonymer:
di-tert-butyl chloromethyl phosphate
Molekylvægt (g/mol):
258.68
Molekylformel:
C9H20ClO4P
InChi Key:
LNJAJHJFSKUCIR-UHFFFAOYSA-N
SMILES:
CC(C)(C)OP(=O)(OCCl)OC(C)(C)C
1
–
3
af
3
Resultater
1
Di-tert-butyl ChloromethylPhosphate (stabilized with K2CO3), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Di-tert-butyl ChloromethylPhosphate (stabilized with K2CO3), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Di-tert-butyl ChloromethylPhosphate (stabilized with K2CO3), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.