Få mere at vide
Di-tert-butyl ChloromethylPhosphate (stabilized with K2CO3), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Di-tert-butyl Chloromethyl Phosphate |
| CAS | 229625-50-7 |
| Molekylær formel | C9 H20 Cl O4 P |
| Mængde | 100 mg |
| Synonym | Solution, contains solid potassium carbonate for buffering purposes, GSK1053539A, Fostemsavir Tromethamine related (GSK3684934A (BMS-663068-03)), Phosphoric acid, chloromethyl bis(1,1-dimethylethyl) ester, Chloromethyl di-tert-butyl phosphate, Di-tert-butyl chloromethyl phosphate, Phosphoric acid di(tert-butyl) chloromethyl ester, Phosphoric acid ditert-butyl ester chloromethyl ester, BMS-371000 |
| InChI formel | InChI=1S/C9H20ClO4P/c1-8(2,3)13-15(11,12-7-10)14-9(4,5)6/h7H2,1-6H3 |
| SMIL | CC(C)(C)OP(=O)(OCCl)OC(C)(C)C |
| IUPAC navn | ditert-butyl chloromethyl phosphate |
| Molekylvægt (g/mol) | 258.6795 |
| Formel vægt | 258.0788 |