CAS RN 22852-13-7
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Tocris Bioscience™ N-Acetylpuromycin
Downregulates SnoN and Ski protein expression; promotes TGF-β signaling
2
Tocris Bioscience™ N-Acetylpuromycin
Downregulates SnoN and Ski protein expression; promotes TGF-β signaling