CAS RN 2246-44-8
IUPAC Navn:
2-methylnaphthalen-1-amine
Synonymer:
1-naphthylamine, 2-methyl-
Molekylvægt (g/mol):
157.22
Molekylformel:
C11H11N
InChi Key:
JMBLSGAXSMOKPN-UHFFFAOYSA-N
SMILES:
CC1=CC=C2C=CC=CC2=C1N
1
–
3
af
3
Resultater
1
1-Amino-2-Methylnaphthalene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Amino-2-Methylnaphthalene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-Amino-2-Methylnaphthalene, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.