CAS RN 2243736-45-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
FB23-2, MedChemExpress
MedChemExpress FB23-2 is a potent and selective inhibitor of mRNA N6-methyladenosine (m6A) demethylase FTO, with an IC50 of 2.6 μM. FB23-2 has anti-proliferation activity. FB23-2 can be used for the research of acute myeloid leukemia (AML).