CAS RN 2239305-70-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
IACS-8968 S-enantiomer, MedChemExpress
MedChemExpress IACS-8968 (S-enantiomer) is the S-enantiomer of IACS-8968. IACS-8968 is a dual IDO and TDO inhibitor, with pIC50s of 6.43 for IDO and <5 for TDO, respectively.