Få mere at vide
IACS-8968 S-enantiomer, MedChemExpress
Artikelnummer. 16464588
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16464588 10 mg
16451169 1 mg
16417307 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16464588 Leverandør Medchem Express Leverandørnr. HY112164B
MedChemExpress IACS-8968 (S-enantiomer) is the S-enantiomer of IACS-8968. IACS-8968 is a dual IDO and TDO inhibitor, with pIC50s of 6.43 for IDO and <5 for TDO, respectively.
IACS-8968 (S-enantiomer) is the S-enantiomer of IACS-8968. IACS-8968 is a dual IDO and TDO inhibitor, with pIC50s of 6.43 for IDO and <5 for TDO, respectively.
Tekniske data
Kemisk navn eller materiale IACS-8968 S-enantiomer
CAS 2239305-70-3
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C17H18F3N5O2
Mængde 10 mg
Synonym IDO/TDO Inhibitor (S-enantiomer)
Opløselighedsinformation DMSO : 50 mg/mL (131.11 mM; Need ultrasonic)
SMIL O=C(N1)N[C@@]2(C(C)(C)CN(C3=CC(C(F)(F)F)=CC4=CN=CN34)CC2)C1=O
Molekylvægt (g/mol) 381.35
Formel vægt 381.35
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death