CAS RN 223104-29-8
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
SEA0400, MedChemExpress
MedChemExpress SEA0400 is a novel and selective inhibitor of the Na+-Ca2+ exchanger (NCX), inhibiting Na+-dependent Ca2+ uptake in cultured neurons, astrocytes, and microglia with IC50s of from 5 to 33 nM.
2
Tocris Bioscience™ SEA 0400
Potent Na+/Ca2+ exchanger (NCX) inhibitor.