CAS RN 2226534-49-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Y06137, MedChemExpress
MedChemExpress Y06137 is a potent and selective BET inhibitor for treatment of castration-resistant prostate cancer (CRPC). Y06137 binds to the BRD4(1) bromodomain with a Kd of 81 nM.