CAS RN 221530-44-5
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(1,2,6,7,-Tetrahydro-8H-indeno[5,4-b]furan-8-ylidene)acetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(1,2,6,7,-Tetrahydro-8H-indeno[5,4-b]furan-8-ylidene)acetonitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.