Få mere at vide
(1,2,6,7,-Tetrahydro-8H-indeno[5,4-b]furan-8-ylidene)acetonitrile, TRC
Artikelnummer. 30421562
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421562 5 mg
30410148 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30421562 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2937305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (1,2,6,7,-Tetrahydro-8H-indeno[5,4-b]furan-8-ylidene)acetoni
CAS 221530-44-5
Molekylær formel C13 H11 N O
Mængde 5 mg
InChI formel InChI=1S/C13H11NO/c14-7-5-10-2-1-9-3-4-12-11(13(9)10)6-8-15-12/h3-5H,1-2,6,8H2/b10-5-
SMIL N#C\C=C/1\CCc2ccc3OCCc3c12
IUPAC navn (2Z)-2-(1,2,6,7-tetrahydrocyclopenta[e][1]benzofuran-8-ylidene)acetonitrile
Molekylvægt (g/mol) 197.23
Formel vægt 197.0841