CAS RN 2200214-93-1
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
SHP099 hydrochloride, MedChemExpress
MedChemExpress SHP099 hydrochloride is a potent, selective and orally available SHP2 inhibitor with an IC50 of 70 nM.