Få mere at vide
SHP099 hydrochloride, MedChemExpress
Artikelnummer. 16484296
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16484296 100 mg -
16445727 5 mg -
16429556 50 mg 50mg
16422988 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16484296 Leverandør Medchem Express Leverandørnr. HY100388A
MedChemExpress SHP099 hydrochloride is a potent, selective and orally available SHP2 inhibitor with an IC50 of 70 nM.
SHP099 hydrochloride is a potent, selective and orally available SHP2 inhibitor with an IC50 of 70 nM.
Tekniske data
Kemisk navn eller materiale SHP099 hydrochloride
CAS 2200214-93-1
Farve Orange
Renhedsgrad noter Research
Molekylær formel C16H20Cl3N5
Mængde 100 mg
Opløselighedsinformation DMSO : 4.1 mg/mL (10.55 mM; Need ultrasonic and warming) ∣H2O : ≥ 2.5 mg/mL (6.43 mM)
SMIL NC1=NC(N2CCC(C)(N)CC2)=CN=C1C3=CC=CC(Cl)=C3Cl.[H]Cl
Molekylvægt (g/mol) 388.72
Formel vægt 388.72
Procent renhed 97.83%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease