CAS RN 21871-47-6
IUPAC Navn:
5-(benzylsulfanyl)-2H-1,2,3,4-tetrazole
Synonymer:
5-(benzylsulfanyl)-2H-1,2,3,4-tetrazole
Molekylvægt (g/mol):
192.24
Molekylformel:
C8H8N4S
InChi Key:
GXGKKIPUFAHZIZ-UHFFFAOYSA-N
SMILES:
C(SC1=NNN=N1)C1=CC=CC=C1
1
–
3
af
3
Resultater
1
5-Benzylthio-1H-tetrazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Benzylthio-1H-tetrazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Benzylthio-1H-tetrazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.