CAS RN 217963-18-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
SCH00013, MedChemExpress
MedChemExpress SCH00013 is a cardiotonic agent that primarily acts via an increase in myofibrillar Ca++ sensitivity, have a significant Ca(2+)sensitizing effect at pH 7.2 to 7.4.