CAS RN 217487-17-7
IUPAC Navn:
3,3-dimethyl-1-[4-(propan-2-yl)phenyl]urea
Synonymer:
tolkan isoproturon arelon ipu stefes isoproturon-50 belgran graminon hytane 500L arelon R IP-flo
Molekylvægt (g/mol):
206.29
Molekylformel:
C12H18N2O
InChi Key:
PUIYMUZLKQOUOZ-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=C(NC(=O)N(C)C)C=C1
1
–
4
af
4
Resultater
2
Isoproturon-d6, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Isoproturon-d6, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.